siRNA Details
Gene Symbol: PALLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166108.1 | NP_001159580.1 | 3 | 1245 | ||
NM_001166109.1 | NP_001159581.1 | 2 | 410 | |||
NM_016081.3 | NP_057165.3 | 3 | 1245 | |||
XM_005262861.4 | XP_005262918.1 | 3 | 1272 | |||
XM_005262866.2 | XP_005262923.1 | 2 | 822 | |||
XM_011531768.2 | XP_011530070.1 | 4 | 1526 | |||
XM_011531769.2 | XP_011530071.1 | 4 | 1526 | |||
XM_011531770.2 | XP_011530072.1 | 4 | 1526 | |||
XM_011531771.2 | XP_011530073.1 | 4 | 1526 | |||
XM_011531772.2 | XP_011530074.1 | 4 | 1526 | |||
XM_011531773.1 | XP_011530075.1 | 4 | 1526 | |||
XM_011531774.1 | XP_011530076.1 | 4 | 1526 | |||
XM_011531775.1 | XP_011530077.1 | 2 | 207 | |||
XM_017007910.1 | XP_016863399.1 | 4 | 1526 | |||
XM_017007911.1 | XP_016863400.1 | 4 | 1526 | |||
GenBank mRNA | AF464873.1 | - | 3 | 1245 | ||
AK095512.1 | - | 2 | 372 | |||
AK096458.1 | - | 2 | 971 | |||
BC140850.1 | - | 3 | 1207 | 9021718 | ||
BC144666.1 | - | 3 | 1207 | 9053197 | ||
DA048169.1 | - | 2 | 410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
