siRNA Details
Gene Symbol: PALLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166109.1 | NP_001159581.1 | 11, 12 | 1572 | ||
NM_001166110.1 | NP_001159582.1 | 4, 5 | 1071 | |||
NM_016081.3 | NP_057165.3 | 12, 13 | 2407 | |||
XM_011531769.2 | XP_011530071.1 | 14, 15 | 3360 | |||
XM_011531774.1 | XP_011530076.1 | 13, 14 | 2688 | |||
XM_017007910.1 | XP_016863399.1 | 14, 15 | 3360 | |||
GenBank mRNA | AB023209.1 | - | 4, 5 | 1035 | ||
AF464873.1 | - | 12, 13 | 2407 | |||
AK095512.1 | - | 11, 12 | 1534 | |||
AY211921.1 | - | 3, 4 | 257 | |||
BC013867.2 | - | 3, 4 | 251 | |||
BC140850.1 | - | 12, 13 | 2369 | 9021718 | ||
DA211938.1 | - | 3, 4 | 400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
