siRNA Details
Gene Symbol: IGHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC001872.1 | - | 6 | 1364 | ||
BC002963.1 | - | 6 | 1344 | |||
BC006180.1 | - | 6 | 1364 | |||
BC009851.2 | - | 6 | 1325 | |||
BC011857.2 | - | 7 | 1344 | |||
BC015760.1 | - | 6 | 1396 | |||
BC017356.2 | - | 6 | 1361 | |||
BC019235.2 | - | 6 | 1363 | |||
BC020240.1 | - | 6 | 1392 | |||
BC073758.1 | - | 6 | 1433 | |||
BC073767.1 | - | 6 | 1352 | |||
BC089412.1 | - | 5 | 1358 | |||
BX161420.1 | - | 3 | 1362 | |||
X17115.1 | - | 7 | 1436 | |||
X58529.1 | - | 4 | 1549 | |||
X67292.1 | - | 3 | 905 | |||
X67301.1 | - | 3 | 905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
