siRNA Details
Gene Symbol: KIAA1683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145304.1 | NP_001138776.1 | 4 | 4353 | ||
NM_001145305.1 | NP_001138777.1 | 4 | 3831 | |||
NM_025249.3 | NP_079525.1 | 4 | 3792 | |||
XM_005260084.1 | XP_005260141.1 | 4 | 4278 | |||
XM_011528324.1 | XP_011526626.1 | 3 | 4299 | |||
GenBank mRNA | AB051470.1 | - | 2 | 5435 | ||
AK057466.1 | - | 4 | 1762 | |||
AK093064.1 | - | 2 | 1761 | |||
AK095765.1 | - | 2 | 3832 | |||
AK095771.1 | - | 1 | 2065 | |||
AK302431.1 | - | 4 | 3744 | |||
AL136867.1 | - | 4 | 3709 | |||
BC034327.1 | - | 1 | 1523 | |||
BC112932.1 | - | 4 | 4279 | 5744104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
