siRNA Details
Gene Symbol: KIAA1683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145304.1 | NP_001138776.1 | 3 | 1381 | ||
NM_001145305.1 | NP_001138777.1 | 3 | 1420 | |||
NM_025249.3 | NP_079525.1 | 3 | 1381 | |||
XM_005260084.1 | XP_005260141.1 | 3 | 1306 | |||
XM_011528324.1 | XP_011526626.1 | 2 | 1327 | |||
GenBank mRNA | AK057466.1 | - | 2 | 1288 | ||
AK302431.1 | - | 3 | 1333 | |||
AL136867.1 | - | 3 | 1298 | |||
BC112932.1 | - | 3 | 1307 | 5744104 | ||
BC127099.1 | - | 2 | 1247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
