siRNA Details
Gene Symbol: OLFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282611.1 | NP_001269540.1 | 3 | 714 | ||
NM_001282612.1 | NP_001269541.1 | 3 | 425 | |||
NM_006334.3 | NP_006325.1 | 3 | 788 | |||
NM_014279.4 | NP_055094.1 | 3 | 802 | |||
GenBank mRNA | AK096304.1 | - | 3 | 425 | ||
AK289719.1 | - | 3 | 515 | |||
AK289921.1 | - | 3 | 397 | |||
AK290478.1 | - | 5 | 502 | |||
BC008763.2 | - | 3 | 524 | 3349741 | ||
BC011741.2 | - | 3 | 524 | 3352603 | ||
BT007146.1 | - | 3 | 308 | |||
BX419781.2 | - | 3 | 719 | |||
BX461034.2 | - | 3 | 770 | |||
D82343.1 | - | 3 | 623 | |||
DB069120.1 | - | 3 | 411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
