siRNA Details
Gene Symbol: CAMSAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015447.3 | NP_056262.3 | 17 | 4525 | ||
XM_005263396.3 | XP_005263453.1 | 18 | 4950 | |||
XM_005263397.1 | XP_005263454.1 | 15 | 4051 | |||
XM_011518272.1 | XP_011516574.1 | 17 | 4219 | |||
XM_011518273.2 | XP_011516575.1 | 16 | 4050 | |||
XM_011518274.1 | XP_011516576.1 | 15 | 4080 | |||
XM_017014301.1 | XP_016869790.1 | 17 | 4806 | |||
XM_017014302.1 | XP_016869791.1 | 17 | 4730 | |||
GenBank mRNA | AJ519841.1 | - | 15 | 4050 | ||
AL834528.1 | - | 7 | 877 | |||
BC010646.1 | - | 7 | 1251 | |||
BC012778.2 | - | 7 | 2525 | |||
BC130580.1 | - | 17 | 4184 | 40146611 | ||
BC144082.1 | - | 17 | 4184 | 9052602 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
