siRNA Details
Gene Symbol: ANKHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017747.2 | NP_060217.1 | 21 | 4036 | ||
GenBank mRNA | AF217646.1 | - | 7 | 1115 | ||
AF521882.1 | - | 21 | 3942 | |||
AK000295.1 | - | 13 | 1559 | |||
AK024056.1 | - | 8 | 1116 | |||
AK074173.1 | - | 22 | 4057 | |||
AY070436.1 | - | 14 | 2366 |
Gene Symbol: ANKHD1-EIF4EBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020690.5 | NP_065741.3 | 21 | 4036 | ||
GenBank mRNA | AF521883.1 | - | 21 | 3942 | ||
BC144623.1 | - | 21 | 3976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
