siRNA Details
Gene Symbol: ANKHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017747.2 | NP_060217.1 | 10 | 1871 | ||
NM_017978.2 | NP_060448.1 | 10 | 1838 | |||
NM_024668.3 | NP_078944.2 | 10 | 1871 | |||
GenBank mRNA | AF258557.1 | - | 10 | 1831 | ||
AF521882.1 | - | 10 | 1777 | |||
AK000295.1 | - | 2 | 144 | |||
AK022041.1 | - | 9 | 1407 | |||
AK074173.1 | - | 10 | 1835 | |||
AY070436.1 | - | 2 | 144 | |||
BC004457.1 | - | 10 | 1816 | |||
BC009420.1 | - | 10 | 1731 | |||
BC009909.2 | - | 10 | 1816 | |||
BC117677.2 | - | 10 | 1797 | |||
BC127127.1 | - | 10 | 1797 | |||
BC150486.2 | - | 10 | 1801 |
Gene Symbol: ANKHD1-EIF4EBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020690.5 | NP_065741.3 | 10 | 1871 | ||
GenBank mRNA | AF521883.1 | - | 10 | 1777 | ||
BC144623.1 | - | 10 | 1811 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
