siRNA Details
Gene Symbol: ZNF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001203250.1 | NP_001190179.1 | 4 | 682 | ||
NM_021143.3 | NP_066966.2 | 4 | 691 | |||
GenBank mRNA | AK023094.1 | - | 4 | 585 | ||
AL080125.1 | - | 4 | 563 | |||
BC036714.2 | - | 4 | 609 | 4846514 | ||
BC045814.1 | - | 4 | 526 | 4830061 |
Gene Symbol: ZNF625-ZNF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037802.1 | 8 | 1048 | |||
GenBank mRNA | AK096560.1 | - | 8 | 1048 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
