siRNA Details
Gene Symbol: RNF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184993.1 | NP_001171922.1 | 8 | 855 | ||
NM_031277.2 | NP_112567.2 | 8 | 855 | |||
XM_006719846.3 | XP_006719909.1 | 8 | 1149 | |||
XM_006719849.2 | XP_006719912.1 | 8 | 866 | |||
XM_006719852.2 | XP_006719915.1 | 7 | 744 | |||
XM_011535152.2 | XP_011533454.1 | 12 | 2774 | |||
XM_011535155.2 | XP_011533457.1 | 11 | 1510 | |||
XM_011535156.2 | XP_011533458.1 | 12 | 2774 | |||
XM_011535157.2 | XP_011533459.1 | 12 | 2774 | |||
XM_011535158.2 | XP_011533460.1 | 12 | 2774 | |||
XM_011535159.2 | XP_011533461.1 | 12 | 2774 | |||
XM_011535160.2 | XP_011533462.1 | 12 | 2774 | |||
XM_011535162.1 | XP_011533464.1 | 4 | 390 | |||
XM_017020676.1 | XP_016876165.1 | 8 | 818 | |||
GenBank mRNA | AF285602.1 | - | 8 | 819 | ||
AF285603.1 | - | 8 | 819 | |||
AK302405.1 | - | 7 | 744 | |||
AK302435.1 | - | 8 | 831 | |||
BC146822.1 | - | 8 | 847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
