siRNA Details
Gene Symbol: RTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265589.1 | NP_001252518.1 | 1 | 83 | ||
NM_001265590.1 | NP_001252519.1 | 1 | 83 | |||
NM_001265591.1 | NP_001252520.1 | 1 | 83 | |||
NM_006054.3 | NP_006045.1 | 1 | 83 | |||
NM_201428.2 | NP_958831.1 | 1 | 83 | |||
NM_201429.2 | NP_958832.1 | 1 | 83 | |||
NM_201430.2 | NP_958833.1 | 1 | 83 | |||
NR_049750.1 | 1 | 83 | ||||
NR_049751.1 | 1 | 83 | ||||
XM_011544730.2 | XP_011543032.1 | 1 | 173 | |||
XM_011544731.2 | XP_011543033.1 | 1 | 218 | |||
XM_017017091.1 | XP_016872580.1 | 1 | 173 | |||
XM_017017092.1 | XP_016872581.1 | 1 | 173 | |||
GenBank mRNA | AB209771.1 | - | 1 | 20 | ||
AF059524.1 | - | 1 | 20 | |||
AF119297.1 | - | 1 | 32 | |||
AK075412.1 | - | 1 | 33 | |||
AK222898.1 | - | 1 | 20 | |||
AK295534.1 | - | 1 | 30 | |||
AK297533.1 | - | 1 | 33 | |||
AK304100.1 | - | 1 | 33 | |||
AK307811.1 | - | 1 | 33 | |||
AK308083.1 | - | 1 | 28 | |||
AK309766.1 | - | 1 | 37 | |||
AY427821.2 | - | 1 | 50 | |||
AY750848.1 | - | 1 | 50 | |||
BC011394.1 | - | 1 | 32 | |||
BC022993.1 | - | 1 | 22 | |||
BC100822.2 | - | 1 | 13 | |||
BC100823.2 | - | 1 | 13 | |||
BC118550.1 | - | 1 | 13 | |||
BC118628.1 | - | 1 | 13 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
