siRNA Details
Gene Symbol: RTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265589.1 | NP_001252518.1 | 9 | 3688 | ||
NM_001265590.1 | NP_001252519.1 | 8 | 3352 | |||
NM_001265591.1 | NP_001252520.1 | 4 | 1044 | |||
NM_006054.3 | NP_006045.1 | 7 | 1300 | |||
NM_201428.2 | NP_958831.1 | 8 | 3631 | |||
NM_201429.2 | NP_958832.1 | 8 | 1357 | |||
NM_201430.2 | NP_958833.1 | 6 | 1241 | |||
NR_049750.1 | 6 | 1161 | ||||
NR_049751.1 | 5 | 1102 | ||||
XM_011544730.2 | XP_011543032.1 | 8 | 3719 | |||
XM_011544731.2 | XP_011543033.1 | 7 | 1433 | |||
GenBank mRNA | AF059524.1 | - | 7 | 1237 | ||
AF119297.1 | - | 7 | 1249 | |||
AK075412.1 | - | 7 | 1249 | |||
AK094965.1 | - | 7 | 2354 | |||
AK122727.1 | - | 7 | 1753 | |||
AK127079.1 | - | 8 | 3327 | |||
AK295361.1 | - | 8 | 1262 | |||
AK295534.1 | - | 7 | 1154 | |||
AK297529.1 | - | 4 | 921 | |||
AK307811.1 | - | 8 | 3578 | |||
AY427821.2 | - | 8 | 3598 | |||
AY750848.1 | - | 9 | 3655 | |||
BC000634.1 | - | 7 | 1208 | |||
BC010556.1 | - | 7 | 1213 | |||
BC011394.1 | - | 7 | 1249 | |||
BC022993.1 | - | 7 | 1239 | |||
BC100822.2 | - | 7 | 1230 | |||
BC100823.2 | - | 7 | 1230 | |||
BC105981.1 | - | 7 | 1181 | |||
BC105982.1 | - | 7 | 1212 | |||
BC118550.1 | - | 7 | 1230 | |||
BC118628.1 | - | 7 | 1230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
