siRNA Details
Gene Symbol: KIDINS220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020738.2 | NP_065789.1 | 19 | 2747 | ||
GenBank mRNA | AB033076.2 | - | 19 | 2722 | ||
AK294832.1 | - | 15 | 2051 | |||
AK296459.1 | - | 19 | 2737 | |||
AL133620.1 | - | 19 | 2725 | |||
BC130610.1 | - | 20 | 2842 | 40146641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
