siRNA Details
Gene Symbol: MYCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293228.1 | NP_001280157.1 | 2 | 801 | ||
NM_001293233.1 | NP_001280162.1 | 2 | 491 | |||
NM_005378.5 | NP_005369.2 | 2 | 491 | |||
XM_017004168.1 | XP_016859657.1 | 1 | 322 | |||
GenBank mRNA | AF320053.1 | - | 2 | 365 | ||
AK298225.1 | - | 2 | 371 | |||
AK299495.1 | - | 2 | 365 | |||
BC002712.2 | - | 1 | 209 | 3627330 | ||
BT007384.1 | - | 1 | 68 | |||
BU167361.1 | - | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
