siRNA Details
Gene Symbol: MYCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293228.1 | NP_001280157.1 | 3 | 1837 | ||
NM_001293231.1 | NP_001280160.1 | 2 | 620 | |||
NM_001293233.1 | NP_001280162.1 | 3 | 1527 | |||
NM_005378.5 | NP_005369.2 | 3 | 1527 | |||
XM_017004168.1 | XP_016859657.1 | 2 | 1358 | |||
GenBank mRNA | AK298225.1 | - | 4 | 820 | ||
AK299495.1 | - | 4 | 1155 | |||
BC002712.2 | - | 2 | 1245 | 3627330 | ||
BT007384.1 | - | 2 | 1104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
