siRNA Details
Gene Symbol: COL4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF258350.1 | - | 3 | 511 | ||
AF400430.1 | - | 3 | 511 | |||
AK025912.1 | - | 16 | 2071 | |||
AK294920.1 | - | 20 | 2398 | |||
AY450357.1 | - | 3 | 511 | |||
AY455978.1 | - | 3 | 511 | |||
BC080644.1 | - | 6 | 918 | |||
BX648393.1 | - | 17 | 2137 | |||
J02760.1 | - | 4 | 616 | |||
M24766.1 | - | 15 | 1849 | |||
X05610.1 | - | 8 | 1207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
