siRNA Details
Gene Symbol: COL4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025912.1 | - | 16 | 2756 | ||
BC080644.1 | - | 6 | 1603 | |||
BX648393.1 | - | 17 | 2824 | |||
X05610.1 | - | 8 | 1893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
