siRNA Details
Gene Symbol: ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000123.3 | NP_000114.2 | 1 | 373 | ||
GenBank mRNA | AF462447.1 | - | 1 | 330 | ||
AK294708.1 | - | 1 | 309 | |||
AK299758.1 | - | 1 | 85 | |||
AK303558.1 | - | 1 | 167 | |||
BC031522.2 | - | 1 | 359 | 3869893 | ||
BX647399.1 | - | 1 | 202 | |||
DA195001.1 | - | 1 | 373 | |||
X69978.1 | - | 1 | 144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
