siRNA Details
Gene Symbol: ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000123.3 | NP_000114.2 | 1 | 238 | ||
GenBank mRNA | AF462447.1 | - | 1 | 195 | ||
AK294708.1 | - | 1 | 174 | |||
AK303558.1 | - | 1 | 32 | |||
BC031522.2 | - | 1 | 224 | 3869893 | ||
BX647399.1 | - | 1 | 67 | |||
DA195001.1 | - | 1 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
