siRNA Details
Gene Symbol: ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000123.3 | NP_000114.2 | 3 | 778 | ||
GenBank mRNA | AF462447.1 | - | 3 | 735 | ||
AK299758.1 | - | 3 | 490 | |||
AK303558.1 | - | 4 | 681 | |||
BC031522.2 | - | 3 | 764 | 3869893 | ||
BX647399.1 | - | 3 | 607 | |||
L20046.1 | - | 3 | 400 | |||
X69978.1 | - | 3 | 549 |
Gene Symbol: BIVM-ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204425.1 | NP_001191354.1 | 11 | 1836 | ||
GenBank mRNA | AB209312.1 | - | 11 | 1628 | ||
LM994822.1 | - | 11 | 1714 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
