siRNA Details
Gene Symbol: TMED7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181836.5 | NP_861974.1 | 3 | 880 | ||
GenBank mRNA | AK027512.1 | - | 3 | 874 | ||
AK074962.1 | - | 3 | 700 | |||
AK304210.1 | - | 3 | 481 | |||
BC019349.2 | - | 3 | 726 | 3625342 | ||
BC107426.1 | - | 2 | 224 |
Gene Symbol: TMED7-TICAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164468.3 | NP_001157940.1 | 3 | 880 | ||
NM_001164469.3 | NP_001157941.1 | 3 | 880 | |||
GenBank mRNA | AK075218.1 | - | 3 | 737 | ||
AY275836.1 | - | 2 | 82 | |||
AY304581.1 | - | 3 | 493 | |||
AY304583.1 | - | 3 | 715 | |||
AY304584.1 | - | 3 | 715 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
