siRNA Details
Gene Symbol: DNAJC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032364.5 | NP_115740.5 | 5 | 1938 | ||
GenBank mRNA | AF141342.1 | - | 2 | 232 | ||
AF351784.1 | - | 3 | 269 | |||
AK055945.1 | - | 4 | 976 | |||
AK123025.1 | - | 4 | 1518 | |||
AK291222.1 | - | 5 | 2173 | |||
AY188447.1 | - | 5 | 1855 | |||
BC080655.1 | - | 4 | 890 | |||
BC117146.1 | - | 5 | 1809 | 40125697 | ||
BC117148.1 | - | 5 | 1809 | 40125699 | ||
EF560741.1 | - | 5 | 1860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
