siRNA Details
Gene Symbol: NHSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144060.1 | NP_001137532.1 | 8 | 4762 | ||
NM_020464.1 | NP_065197.1 | 7 | 5242 | |||
XM_005267062.4 | XP_005267119.1 | 8 | 7026 | |||
XM_011535965.2 | XP_011534267.1 | 9 | 7080 | |||
XM_011535966.2 | XP_011534268.1 | 10 | 6756 | |||
XM_011535967.2 | XP_011534269.1 | 10 | 6700 | |||
XM_011535968.2 | XP_011534270.1 | 8 | 6948 | |||
XM_011535969.2 | XP_011534271.1 | 9 | 6406 | |||
XM_011535970.2 | XP_011534272.1 | 9 | 7270 | |||
XM_011535971.2 | XP_011534273.1 | 8 | 5537 | |||
XM_011535972.2 | XP_011534274.1 | 9 | 5351 | |||
XM_011535973.2 | XP_011534275.1 | 8 | 5703 | |||
XM_011535974.2 | XP_011534276.1 | 9 | 6495 | |||
XM_011535976.1 | XP_011534278.1 | 9 | 4747 | |||
XM_017011086.1 | XP_016866575.1 | 10 | 6666 | |||
XM_017011087.1 | XP_016866576.1 | 9 | 5700 | |||
XM_017011088.1 | XP_016866577.1 | 7 | 6294 | |||
XM_017011089.1 | XP_016866578.1 | 8 | 5295 | |||
GenBank mRNA | AB037778.1 | - | 3 | 2302 | ||
AK000699.1 | - | 2 | 1633 | |||
AK025199.1 | - | 5 | 1852 | |||
AK025347.1 | - | 3 | 2266 | |||
AK299585.1 | - | 3 | 1157 | |||
BC045181.1 | - | 1 | 164 | |||
BX647246.1 | - | 4 | 4135 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
