siRNA Details
Gene Symbol: NHSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144060.1 | NP_001137532.1 | 8 | 5772 | ||
NM_020464.1 | NP_065197.1 | 7 | 6252 | |||
XM_005267062.4 | XP_005267119.1 | 8 | 8036 | |||
XM_011535965.2 | XP_011534267.1 | 9 | 8090 | |||
XM_011535966.2 | XP_011534268.1 | 10 | 7766 | |||
XM_011535967.2 | XP_011534269.1 | 10 | 7710 | |||
XM_011535968.2 | XP_011534270.1 | 8 | 7958 | |||
XM_011535969.2 | XP_011534271.1 | 9 | 7416 | |||
XM_011535970.2 | XP_011534272.1 | 9 | 8280 | |||
XM_011535971.2 | XP_011534273.1 | 8 | 6547 | |||
XM_011535972.2 | XP_011534274.1 | 9 | 6361 | |||
XM_011535973.2 | XP_011534275.1 | 8 | 6713 | |||
XM_011535974.2 | XP_011534276.1 | 9 | 7505 | |||
XM_011535976.1 | XP_011534278.1 | 9 | 5757 | |||
XM_017011086.1 | XP_016866575.1 | 10 | 7676 | |||
XM_017011087.1 | XP_016866576.1 | 9 | 6710 | |||
XM_017011088.1 | XP_016866577.1 | 7 | 7304 | |||
XM_017011089.1 | XP_016866578.1 | 8 | 6305 | |||
GenBank mRNA | AB037778.1 | - | 3 | 3312 | ||
AK025199.1 | - | 5 | 2861 | |||
AK025347.1 | - | 3 | 3277 | |||
BC020940.1 | - | 1 | 462 | |||
BC045181.1 | - | 1 | 1174 | |||
BX647246.1 | - | 4 | 5143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
