siRNA Details
Gene Symbol: NHSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144060.1 | NP_001137532.1 | 6 | 2506 | ||
NM_020464.1 | NP_065197.1 | 5 | 2986 | |||
XM_005267062.4 | XP_005267119.1 | 6 | 4770 | |||
XM_011535965.2 | XP_011534267.1 | 7 | 4824 | |||
XM_011535966.2 | XP_011534268.1 | 8 | 4500 | |||
XM_011535967.2 | XP_011534269.1 | 8 | 4444 | |||
XM_011535968.2 | XP_011534270.1 | 6 | 4692 | |||
XM_011535969.2 | XP_011534271.1 | 7 | 4150 | |||
XM_011535970.2 | XP_011534272.1 | 7 | 5014 | |||
XM_011535971.2 | XP_011534273.1 | 6 | 3281 | |||
XM_011535972.2 | XP_011534274.1 | 7 | 3095 | |||
XM_011535973.2 | XP_011534275.1 | 6 | 3447 | |||
XM_011535974.2 | XP_011534276.1 | 7 | 4239 | |||
XM_011535976.1 | XP_011534278.1 | 7 | 2491 | |||
XM_017011086.1 | XP_016866575.1 | 8 | 4410 | |||
XM_017011087.1 | XP_016866576.1 | 7 | 3444 | |||
XM_017011088.1 | XP_016866577.1 | 5 | 4038 | |||
XM_017011089.1 | XP_016866578.1 | 6 | 3039 | |||
GenBank mRNA | AB037778.1 | - | 1 | 46 | ||
AK307584.1 | - | 6 | 2506 | |||
BX647246.1 | - | 2 | 1879 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
