siRNA Details
Gene Symbol: CCDC142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032779.3 | NP_116168.3 | 4, 5 | 1753 | ||
GenBank mRNA | AK075543.1 | - | 4, 5 | 1519 | ||
BC117269.1 | - | 4, 5 | 1471 | 40125820 | ||
BC143398.1 | - | 4, 5 | 1471 | 9051905 | ||
BC143399.1 | - | 4, 5 | 1471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
