siRNA Details
Gene Symbol: CCDC142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032779.3 | NP_116168.3 | 7 | 2114 | ||
GenBank mRNA | AK027303.1 | - | 2 | 199 | ||
AK075543.1 | - | 7 | 1880 | |||
BC117269.1 | - | 7 | 1832 | 40125820 | ||
BC143398.1 | - | 7 | 1832 | 9051905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
