siRNA Details
Gene Symbol: CSHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318.3 | NP_001309.3 | 2, 3 | 272 | ||
NM_001321067.1 | NP_001307996.1 | 2, 3 | 268 | |||
NM_001321068.1 | NP_001307997.1 | 3, 4 | 262 | |||
NM_001321069.1 | NP_001307998.1 | 3, 4 | 331 | |||
NM_022579.2 | NP_072101.1 | 3, 4 | 448 | |||
NM_022580.2 | NP_072102.1 | 2, 3 | 199 | |||
NM_022581.2 | NP_072103.1 | 3, 4 | 379 | |||
XM_011524344.1 | XP_011522646.1 | 3, 4 | 378 | |||
XM_011524346.2 | XP_011522648.1 | 2, 3 | 220 | |||
GenBank mRNA | AK300190.1 | - | 2, 3 | 587 | ||
AX772828.1 | - | 3, 4 | 276 | |||
BC029365.1 | - | 2, 3 | 202 | 4619624 | ||
BC119747.2 | - | 2, 3 | 199 | 40119241 | ||
BX382469.2 | - | 2, 3 | 272 | |||
BX426214.2 | - | 3, 4 | 244 | |||
DR004701.1 | - | 3, 4 | 172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
