siRNA Details
Gene Symbol: TRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243879.1 | NP_001230808.1 | 1 | 234 | ||
NM_004593.2 | NP_004584.1 | 1 | 234 | |||
XM_005247703.1 | XP_005247760.1 | 1 | 126 | |||
GenBank mRNA | AK098191.1 | - | 1 | 129 | ||
AK301118.1 | - | 1 | 87 | |||
AK308267.1 | - | 1 | 112 | |||
AK315632.1 | - | 1 | 218 | |||
BC000160.2 | - | 1 | 101 | 2984771 | ||
BC000451.2 | - | 1 | 100 | 2821443 | ||
BC005898.1 | - | 1 | 85 | 4046349 | ||
DA164444.1 | - | 1 | 234 | |||
U61267.1 | - | 1 | 79 | |||
U68063.1 | - | 1 | 87 | |||
U87836.1 | - | 1 | 116 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
