siRNA Details
Gene Symbol: TRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243879.1 | NP_001230808.1 | 8 | 1118 | ||
NM_004593.2 | NP_004584.1 | 9 | 1252 | |||
XM_005247703.1 | XP_005247760.1 | 10 | 1420 | |||
XM_011513072.2 | XP_011511374.1 | 10 | 4222 | |||
XM_017007033.1 | XP_016862522.1 | 9 | 2398 | |||
GenBank mRNA | AB209208.1 | - | 10 | 4208 | ||
AK091863.1 | - | 5 | 2139 | |||
AK094495.1 | - | 6 | 1174 | |||
AK098191.1 | - | 9 | 1039 | |||
AK125610.1 | - | 6 | 1922 | |||
AK301118.1 | - | 8 | 971 | |||
AK308267.1 | - | 6 | 837 | |||
BC000160.2 | - | 9 | 1119 | 2984771 | ||
BC000451.2 | - | 9 | 1118 | 2821443 | ||
BC005898.1 | - | 9 | 1103 | 4046349 | ||
CR749214.1 | - | 8 | 3279 | |||
U61267.1 | - | 9 | 1097 | |||
U68063.1 | - | 9 | 1105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
