siRNA Details
Gene Symbol: CNPY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318842.1 | NP_001305771.1 | 4 | 898 | ||
NM_001318845.1 | NP_001305774.1 | 2 | 675 | |||
NM_001318847.1 | NP_001305776.1 | 4 | 861 | |||
NM_001318848.1 | NP_001305777.1 | 3 | 799 | |||
NM_006586.4 | NP_006577.2 | 3 | 799 | |||
NR_134880.1 | 4 | 861 | ||||
NR_134881.1 | 5 | 2054 | ||||
NR_134882.1 | 3 | 737 | ||||
NR_134885.1 | 3 | 896 | ||||
NR_134886.1 | 2 | 675 | ||||
NR_134888.1 | 2 | 675 | ||||
GenBank mRNA | AK090425.1 | - | 1 | 1223 | ||
AK124196.1 | - | 5 | 2053 | |||
AY358960.1 | - | 3 | 694 | |||
BC004423.1 | - | 3 | 745 | 2819660 | ||
BC008133.1 | - | 2 | 311 | |||
BC008898.2 | - | 3 | 426 | 2959532 | ||
BC008961.2 | - | 3 | 426 | 2959532 | ||
BG675987.1 | - | 4 | 455 | |||
BI909527.1 | - | 3 | 389 | |||
BQ063472.1 | - | 4 | 504 | |||
BQ680146.1 | - | 3 | 418 | |||
CK429647.1 | - | 4 | 474 | |||
CN302712.1 | - | 1 | 42 | |||
CN302713.1 | - | 3 | 537 | |||
HY101691.1 | - | 2 | 323 | |||
U80744.1 | - | 3 | 744 |
Gene Symbol: CNPY3-GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134890.1 | 2 | 675 | |||
GenBank mRNA | FY213816.1 | - | 2 | 207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
