siRNA Details
Gene Symbol: CNPY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318842.1 | NP_001305771.1 | 1 | 451 | ||
NM_001318845.1 | NP_001305774.1 | 1 | 451 | |||
NM_001318847.1 | NP_001305776.1 | 1 | 451 | |||
NM_001318848.1 | NP_001305777.1 | 1 | 451 | |||
NM_006586.4 | NP_006577.2 | 1 | 451 | |||
NR_134880.1 | 1 | 451 | ||||
NR_134881.1 | 1 | 1579 | ||||
NR_134882.1 | 1 | 451 | ||||
NR_134885.1 | 1 | 451 | ||||
NR_134886.1 | 1 | 451 | ||||
NR_134888.1 | 1 | 451 | ||||
GenBank mRNA | AK124196.1 | - | 1 | 1578 | ||
AY358960.1 | - | 1 | 346 | |||
BC004423.1 | - | 1 | 397 | 2819660 | ||
BC008133.1 | - | 1 | 87 | |||
BC008898.2 | - | 1 | 78 | 2959532 | ||
BC008961.2 | - | 1 | 78 | 2959532 | ||
BF528267.1 | - | 1 | 43 | |||
BG675987.1 | - | 1 | 45 | |||
BI909527.1 | - | 1 | 103 | |||
BQ063472.1 | - | 1 | 57 | |||
BQ680146.1 | - | 1 | 70 | |||
CK429647.1 | - | 1 | 64 | |||
CN302713.1 | - | 1 | 92 | |||
HY101691.1 | - | 1 | 99 | |||
U80744.1 | - | 1 | 397 |
Gene Symbol: CNPY3-GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318856.1 | NP_001305785.1 | 1 | 451 | ||
NM_001318857.1 | NP_001305786.1 | 1 | 451 | |||
NM_001318858.1 | NP_001305787.1 | 1 | 451 | |||
NR_134890.1 | 1 | 451 | ||||
NR_134891.1 | 1 | 451 | ||||
NR_134892.1 | 1 | 451 | ||||
GenBank mRNA | BQ947041.1 | - | 1 | 78 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
