siRNA Details
Gene Symbol: MOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008228.2 | NP_001008229.1 | 1 | 250 | ||
NM_001008229.2 | NP_001008230.1 | 1 | 250 | |||
NM_001170418.1 | NP_001163889.1 | 1 | 250 | |||
NM_002433.4 | NP_002424.3 | 1 | 250 | |||
NM_206809.3 | NP_996532.2 | 1 | 250 | |||
NM_206810.3 | NP_996533.2 | 1 | 250 | |||
NM_206811.3 | NP_996534.2 | 1 | 250 | |||
NM_206812.3 | NP_996535.2 | 1 | 250 | |||
NM_206814.5 | NP_996537.3 | 1 | 250 | |||
XM_005249131.3 | XP_005249188.1 | 1 | 239 | |||
GenBank mRNA | BX107434.1 | - | 1 | 95 | ||
DA139582.1 | - | 1 | 241 | |||
DA140757.1 | - | 1 | 100 | |||
U64564.1 | - | 1 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
