siRNA Details
Gene Symbol: MOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008228.2 | NP_001008229.1 | 4, 5 | 799 | ||
NM_001008229.2 | NP_001008230.1 | 4, 5 | 799 | |||
NM_001170418.1 | NP_001163889.1 | 3, 4 | 451 | |||
NM_002433.4 | NP_002424.3 | 4, 5 | 799 | |||
NM_206809.3 | NP_996532.2 | 4, 5 | 799 | |||
NM_206810.3 | NP_996533.2 | 4, 5 | 799 | |||
NM_206811.3 | NP_996534.2 | 4, 5 | 799 | |||
NM_206812.3 | NP_996535.2 | 4, 5 | 799 | |||
NM_206814.5 | NP_996537.3 | 3, 4 | 451 | |||
XM_005249131.3 | XP_005249188.1 | 4, 5 | 788 | |||
GenBank mRNA | AB209815.1 | - | 4, 5 | 709 | ||
AK295213.1 | - | 5, 6 | 620 | |||
AK308949.1 | - | 3, 4 | 429 | |||
AK312892.1 | - | 4, 5 | 685 | |||
AY566846.1 | - | 3, 4 | 216 | |||
AY566847.1 | - | 3, 4 | 216 | |||
AY566852.1 | - | 4, 5 | 570 | |||
AY566853.1 | - | 4, 5 | 564 | |||
AY566854.1 | - | 4, 5 | 564 | |||
BC035938.1 | - | 4, 5 | 688 | 4791512 | ||
BX107434.1 | - | 4, 5 | 645 | |||
U18798.1 | - | 4, 5 | 570 | |||
U18799.1 | - | 4, 5 | 570 | |||
U18800.2 | - | 3, 4 | 222 | |||
U18801.1 | - | 4, 5 | 570 | |||
U18803.1 | - | 4, 5 | 570 | |||
U18840.1 | - | 4, 5 | 570 | |||
U18843.1 | - | 4, 5 | 570 | |||
U64564.1 | - | 4, 5 | 732 | |||
U64565.1 | - | 3, 4 | 483 | |||
U64566.1 | - | 3, 4 | 483 | |||
U64567.1 | - | 3, 4 | 483 | |||
U64568.1 | - | 3, 4 | 483 | |||
U64569.1 | - | 3, 4 | 483 | |||
U64570.1 | - | 4, 5 | 621 | |||
U64571.1 | - | 4, 5 | 621 | |||
X74511.1 | - | 4, 5 | 570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
