siRNA Details
Gene Symbol: MOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008228.2 | NP_001008229.1 | 2 | 330 | ||
NM_001008229.2 | NP_001008230.1 | 2 | 330 | |||
NM_002433.4 | NP_002424.3 | 2 | 330 | |||
NM_206809.3 | NP_996532.2 | 2 | 330 | |||
NM_206810.3 | NP_996533.2 | 2 | 330 | |||
NM_206811.3 | NP_996534.2 | 2 | 330 | |||
NM_206812.3 | NP_996535.2 | 2 | 330 | |||
XM_005249131.3 | XP_005249188.1 | 2 | 319 | |||
GenBank mRNA | AB209815.1 | - | 2 | 240 | ||
AK295213.1 | - | 2 | 206 | |||
AK312892.1 | - | 2 | 216 | |||
AY566848.1 | - | 2 | 95 | |||
AY566851.1 | - | 2 | 95 | |||
AY566852.1 | - | 2 | 101 | |||
AY566853.1 | - | 2 | 95 | |||
AY566854.1 | - | 2 | 95 | |||
BC035938.1 | - | 2 | 219 | 4791512 | ||
BG699861.1 | - | 1, 2 | 264 | |||
BI545470.1 | - | 2 | 317 | |||
BX107434.1 | - | 2 | 175 | |||
DA139582.1 | - | 2 | 321 | |||
DA140757.1 | - | 2 | 180 | |||
DA252279.1 | - | 2 | 219 | |||
U18798.1 | - | 2 | 101 | |||
U18799.1 | - | 2 | 101 | |||
U18801.1 | - | 2 | 101 | |||
U18803.1 | - | 2 | 101 | |||
U18840.1 | - | 2 | 101 | |||
U18843.1 | - | 2 | 101 | |||
U64564.1 | - | 2 | 263 | |||
U64565.1 | - | 1 | 14 | |||
U64566.1 | - | 1 | 14 | |||
U64567.1 | - | 1 | 14 | |||
U64568.1 | - | 1 | 14 | |||
U64569.1 | - | 1 | 14 | |||
U64570.1 | - | 1 | 14 | |||
U64571.1 | - | 1 | 14 | |||
X74511.1 | - | 2 | 101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
