siRNA Details
Gene Symbol: SLTM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013843.2 | NP_001013865.1 | 11 | 1492 | ||
NM_024755.3 | NP_079031.2 | 11 | 1546 | |||
NR_135042.1 | 9 | 879 | ||||
NR_135043.1 | 9 | 989 | ||||
XM_006720686.3 | XP_006720749.3 | 9 | 1937 | |||
XM_006720690.1 | XP_006720753.1 | 7 | 643 | |||
XM_011522022.1 | XP_011520324.1 | 11 | 2204 | |||
XM_011522023.1 | XP_011520325.1 | 11 | 2150 | |||
XM_011522024.1 | XP_011520326.1 | 10 | 2054 | |||
XM_011522025.1 | XP_011520327.1 | 10 | 2006 | |||
XM_011522026.2 | XP_011520328.2 | 10 | 1985 | |||
XM_011522027.1 | XP_011520329.1 | 9 | 1958 | |||
XM_011522028.1 | XP_011520330.1 | 9 | 1904 | |||
XM_011522029.2 | XP_011520331.1 | 11 | 1385 | |||
XM_011522030.2 | XP_011520332.1 | 11 | 2204 | |||
XM_011522031.2 | XP_011520333.1 | 8 | 1040 | |||
XM_017022576.1 | XP_016878065.1 | 10 | 1342 | |||
XM_017022577.1 | XP_016878066.1 | 10 | 1931 | |||
XM_017022578.1 | XP_016878067.1 | 9 | 1883 | |||
XM_017022579.1 | XP_016878068.1 | 11 | 1549 | |||
XM_017022580.1 | XP_016878069.1 | 10 | 1003 | |||
GenBank mRNA | AK000867.1 | - | 11 | 1438 | ||
AK024710.1 | - | 5 | 859 | |||
AK055195.1 | - | 9 | 782 | |||
AK291423.1 | - | 9 | 912 | |||
BC014944.1 | - | 4 | 349 | |||
BC140851.1 | - | 11 | 1513 | 9021719 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
