siRNA Details
Gene Symbol: KNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144508.4 | NP_653091.3 | 6 | 382 | ||
NM_170589.4 | NP_733468.3 | 6 | 382 | |||
XM_017022432.1 | XP_016877921.1 | 6 | 619 | |||
GenBank mRNA | AB022190.1 | - | 6 | 383 | ||
AF173994.1 | - | 6 | 611 | |||
AF248041.1 | - | 6 | 366 | |||
AF461041.1 | - | 6 | 366 | |||
AK307996.1 | - | 6 | 372 | |||
BC029373.1 | - | 6 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
