siRNA Details
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162536.2 | NP_001156008.1 | 1 | 336 | ||
NM_019610.5 | NP_062556.2 | 1 | 336 | |||
GenBank mRNA | AL832554.1 | - | 1 | 188 | ||
DA010727.1 | - | 1 | 327 | |||
DA216732.1 | - | 1 | 178 | |||
DA390224.1 | - | 1 | 152 | |||
DC397531.1 | - | 1 | 403 |
Gene Symbol: KYAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008661.2 | NP_001008661.1 | 1 | 336 | ||
NM_001008662.2 | NP_001008662.1 | 1 | 336 | |||
GenBank mRNA | AK057176.1 | - | 1 | 152 | ||
AK301326.1 | - | 1 | 23 | |||
AK315917.1 | - | 1 | 33 | |||
AY028624.1 | - | 1 | 156 | |||
BM728982.1 | - | 1 | 298 | |||
DC424120.1 | - | 1 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
