siRNA Details
Gene Symbol: KYAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008661.2 | NP_001008661.1 | 1 | 362 | ||
NM_001008662.2 | NP_001008662.1 | 1 | 362 | |||
GenBank mRNA | AK057176.1 | - | 1 | 178 | ||
AK301326.1 | - | 1 | 49 | |||
AK315917.1 | - | 1 | 59 | |||
AY028624.1 | - | 1 | 182 | |||
BM728982.1 | - | 1 | 324 | |||
CR627392.1 | - | 1 | 24 | |||
DC424120.1 | - | 1 | 365 |
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162536.2 | NP_001156008.1 | 1 | 362 | ||
NM_019610.5 | NP_062556.2 | 1 | 362 | |||
GenBank mRNA | AL832554.1 | - | 1 | 214 | ||
BC012942.1 | - | 1 | 21 | 3875565 | ||
DA010727.1 | - | 1 | 353 | |||
DA216732.1 | - | 1 | 204 | |||
DA390224.1 | - | 1 | 178 | |||
DC397531.1 | - | 1 | 429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
