siRNA Details
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162536.2 | NP_001156008.1 | 3 | 2427 | ||
NM_019610.5 | NP_062556.2 | 2 | 2327 | |||
GenBank mRNA | AL832554.1 | - | 2 | 2180 | ||
AL832785.1 | - | 1 | 1271 | |||
BC012942.1 | - | 2 | 1986 | 3875565 |
Gene Symbol: RBMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647131.1 | - | 7 | 2320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
