siRNA Details
Gene Symbol: CUZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022034.5 | NP_071317.2 | 7 | 1548 | ||
NR_037912.1 | 6 | 1397 | ||||
GenBank mRNA | AF305835.1 | - | 4 | 1335 | ||
AK289445.1 | - | 6 | 1079 | |||
AY260047.1 | - | 7 | 1548 | |||
AY260048.1 | - | 6 | 1248 | |||
AY260049.1 | - | 6 | 1397 | |||
AY260050.1 | - | 2 | 640 | |||
BC136755.1 | - | 7 | 1386 | 9020745 | ||
BC144398.1 | - | 7 | 1237 | |||
BX538283.1 | - | 4 | 872 | |||
BX538284.1 | - | 4 | 925 | |||
BX648572.1 | - | 7 | 1230 |
Gene Symbol: FAM24B-CUZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037915.1 | 9 | 1892 | |||
GenBank mRNA | AK300849.1 | - | 9 | 1881 | ||
AY358556.1 | - | 9 | 2179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
