siRNA Details
Gene Symbol: FAM24B-CUZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037915.1 | 10 | 2127 | |||
GenBank mRNA | AK300849.1 | - | 10 | 2116 | ||
AY358556.1 | - | 10 | 2414 |
Gene Symbol: CUZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022034.5 | NP_071317.2 | 8 | 1783 | ||
NR_037912.1 | 7 | 1632 | ||||
GenBank mRNA | AF305835.1 | - | 5 | 1570 | ||
AK289445.1 | - | 7 | 1314 | |||
AY260047.1 | - | 8 | 1783 | |||
AY260048.1 | - | 7 | 1483 | |||
AY260049.1 | - | 7 | 1632 | |||
AY260050.1 | - | 3 | 875 | |||
BC136755.1 | - | 8 | 1621 | 9020745 | ||
BC144398.1 | - | 8 | 1472 | |||
BX538283.1 | - | 5 | 1107 | |||
BX538284.1 | - | 5 | 1160 | |||
BX648572.1 | - | 8 | 1465 |

More Information
Other Species Targeted

Gene Ontology Categories:
