siRNA Details
Gene Symbol: ZNF365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014951.2 | NP_055766.2 | 2 | 344 | ||
NM_199450.2 | NP_955522.1 | 2 | 344 | |||
NM_199451.2 | NP_955523.1 | 2 | 344 | |||
XM_017015937.1 | XP_016871426.1 | 2 | 892 | |||
GenBank mRNA | AB020651.1 | - | 2 | 306 | ||
AJ505147.1 | - | 2 | 117 | |||
AJ505148.1 | - | 2 | 306 | |||
AJ505149.1 | - | 2 | 306 | |||
AK289515.1 | - | 2 | 85 | |||
AK289808.1 | - | 2 | 109 | |||
BC017841.1 | - | 2 | 148 | |||
BC070073.1 | - | 2 | 129 | |||
BX647853.1 | - | 2 | 137 | |||
BX647904.1 | - | 2 | 137 | |||
DB476702.1 | - | 2 | 341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
