siRNA Details
Gene Symbol: FRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166133.1 | NP_001159605.1 | 13 | 1714 | ||
NM_025074.6 | NP_079350.5 | 13 | 1714 | |||
XM_006714316.2 | XP_006714379.1 | 13 | 1961 | |||
GenBank mRNA | AK027833.1 | - | 8 | 699 | ||
BC020192.1 | - | 5 | 494 | |||
BC052281.1 | - | 8 | 801 | |||
BC064487.1 | - | 8 | 801 | |||
BC094853.1 | - | 5 | 494 | |||
BC131820.1 | - | 13 | 1304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
