siRNA Details
Gene Symbol: FRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166133.1 | NP_001159605.1 | 8 | 1167 | ||
NM_025074.6 | NP_079350.5 | 8 | 1167 | |||
XM_006714316.2 | XP_006714379.1 | 8 | 1414 | |||
GenBank mRNA | AK027833.1 | - | 3 | 152 | ||
BC052281.1 | - | 3 | 254 | |||
BC064487.1 | - | 3 | 254 | |||
BC131820.1 | - | 8 | 757 | |||
BC131821.1 | - | 8 | 757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
