siRNA Details
Gene Symbol: HNRNPA1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011724.1 | NP_001011724.1 | 1 | 196 | ||
NM_001011725.1 | NP_001011725.1 | 1 | 196 | |||
GenBank mRNA | AK123159.1 | - | 1 | 152 | ||
BC082242.1 | - | 1 | 127 | |||
BQ425201.1 | - | 1 | 164 | |||
BX477483.1 | - | 1 | 198 | |||
CD523364.1 | - | 1 | 167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
