siRNA Details
Gene Symbol: HNRNPA1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011724.1 | NP_001011724.1 | 7 | 1288 | ||
NM_001011725.1 | NP_001011725.1 | 6 | 1147 | |||
GenBank mRNA | AK091082.1 | - | 1 | 1000 | ||
AK094423.1 | - | 6 | 1038 | |||
AK123159.1 | - | 5 | 827 | |||
AK126454.1 | - | 1 | 1922 | |||
BC082242.1 | - | 6 | 1078 | |||
BC108266.1 | - | 1 | 276 | 4778431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
