siRNA Details
Gene Symbol: HYPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199885.1 | NP_001186814.1 | 3 | 535 | ||
NM_016400.3 | NP_057484.3 | 4 | 591 | |||
GenBank mRNA | AF161485.1 | - | 4 | 412 | ||
AK022435.1 | - | 2 | 498 | |||
BC019262.1 | - | 4 | 589 | 3504222 | ||
BQ478474.1 | - | 3 | 333 |
Gene Symbol: SERF2-C15ORF63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037673.1 | 6 | 1034 | |||
GenBank mRNA | AF370428.1 | - | 5 | 4527 | ||
AK000438.1 | - | 6 | 656 | |||
AK094716.1 | - | 5 | 2359 | |||
AK095876.1 | - | 7 | 2032 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
