siRNA Details
Gene Symbol: SERF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018108.3 | NP_001018118.1 | 2 | 557 | ||
NM_001199875.1 | NP_001186804.1 | 2 | 423 | |||
NM_001199876.1 | NP_001186805.1 | 2 | 423 | |||
NM_001199877.1 | NP_001186806.1 | 4 | 955 | |||
NM_001199878.1 | NP_001186807.1 | 2 | 250 | |||
NR_037672.1 | 2 | 423 | ||||
GenBank mRNA | AF073298.1 | - | 2 | 81 | ||
AF320073.1 | - | 2 | 75 | |||
AK000479.1 | - | 2 | 69 | |||
AV713454.2 | - | 2 | 120 | |||
BC008214.1 | - | 2 | 557 | |||
BC009869.2 | - | 4 | 953 | |||
BC015491.1 | - | 2 | 54 | 3902770 | ||
BC022326.1 | - | 2 | 72 | 4134511 | ||
BC038893.1 | - | 2 | 38 | |||
BI260545.1 | - | 2 | 157 | |||
BP317864.2 | - | 2 | 77 | |||
DA302996.1 | - | 2 | 251 | |||
DB060650.1 | - | 2 | 79 |
Gene Symbol: SERF2-C15ORF63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037673.1 | 2 | 423 | |||
GenBank mRNA | AK000438.1 | - | 2 | 45 | ||
AK095876.1 | - | 2 | 45 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
